3-Phenyl-1-propyne ≥97%

Synonyms: 2-Propynylbenzene

376841-1G 376841-5G
220061-935EA 109.85 USD
220061-935 220061-936
3-Phenyl-1-propyne ≥97%
3-Phenyl-1-propyne
Formula: C₉H₈
MW: 116.16 g/mol
MDL Number: MFCD00134431
CAS Number: 10147-11-2
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR