1,2-Bis(diphenylphosphino)ethane

Warning

Synonyms: Ethylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)ethane, DIPHOS, 1,2-Bis(diphenylphosphino)ethane, Ethane-1,3-diylbis(diphenylphosphine)

376728-1G 376728-5G
220062-269EA 23.54 USD
220062-269 220062-270
1,2-Bis(diphenylphosphino)ethane
1,2-Bis(diphenylphosphino)ethane
Formula: C₂₆H₂₄P₂
MW: 398.42 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00003047
CAS Number: 1663-45-2
EINECS: 216-769-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR