2,8-Quinolinediol

Warning

Synonyms: 2,8-Dihydroxyquinoline

37640-1G-F 37640-5G-F
220062-239EA 44.8 USD
220062-239 220062-241
2,8-Quinolinediol
2,8-Quinolinediol
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00216696
CAS Number: 15450-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR