Octadecyltrimethoxysilane, Technical Grade

Synonyms: Trimethoxy(octadecyl)silane

376213-5ML 376213-25ML
220062-229EA 67.74 USD
220062-229 220062-228
Octadecyltrimethoxysilane, Technical Grade
Octadecyltrimethoxysilane
Formula: C₂₁H₄₆O₃Si
MW: 374.68 g/mol
MDL Number: MFCD00043060
CAS Number: 3069-42-9
EINECS: 221-339-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR