3,4-Dihydroxybenzoic acid

Synonyms: Protocatechuic acid

37580-100G-F 37580-25G-F
220061-159EA 228.31 USD
220061-159 220062-206
3,4-Dihydroxybenzoic acid
3,4-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 125 - 135 °C (0.001 -rr)
Melting Pt: 202 - 204 °C
Density: 1.5415 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002509
CAS Number: 99-50-3
EINECS: 202-760-0
Merck Index: 12,08080

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR