2-Methyl-1,3-propanediol ≥99%

Synonyms: 1,3-Dihydroxy-2-methylpropane

375721-3L 375721-1L
220061-151EA 159.54 USD
220061-151 220061-150
2-Methyl-1,3-propanediol ≥99%
2-Methyl-1,3-propanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
MDL Number: MFCD00082586
CAS Number: 2163-42-0
EINECS: 412-350-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR