Dilithium oxide ≥97%

Danger

Synonyms: Lithium oxide

374725-10G 374725-100G
220062-027EA 156.92 USD
220062-027 220062-026
Dilithium oxide ≥97%
di-Lithium oxide
Formula: Li₂O
MW: 29.88 g/mol
Melting Pt: 1450 °C
Density: 2.013 g/cm³ (25 °C)
MDL Number: MFCD00016183
CAS Number: 12057-24-8
EINECS: 235-019-5
UN: 3262
ADR: 8,II
Merck Index: 13,05560

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR