Tributylphenyltin

Danger

Synonyms: Tri-n-butylphenyltin, Tributylphenylstannane

374601-10G 374601-1G
220059-750EA 126.69 USD
220059-750 220059-751
Tributylphenyltin
Tributylphenyltin
Formula: C₁₈H₃₂Sn
MW: 367.16 g/mol
MDL Number: MFCD00134394
CAS Number: 960-16-7
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR