(S)-(-)-Citronellal ≥96%

Synonyms: (S)-(-)-3,7-Dimethyl-6-octenal, (S)-(-)-3,7-Dimethyloct-6-enal, (-)-Citronellal, (S)-3,7-Dimethyloct-6-enal

373753-5G 373753-1G
220061-870EA 143.23 USD
220061-870 220061-869
(S)-(-)-Citronellal ≥96%
(S)-(-)-Citronellal
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 84 °C (1013 hPa)
Melting Pt: -63 °C
Density: 0.85 g/cm³ (20 °C)
MDL Number: MFCD00075588
CAS Number: 5949-05-3
EINECS: 227-707-9

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR