Sodium perborate monohydrate

Danger

372862-100G 372862-500G
220061-817EA 101.23 USD
220061-817 220061-818
Sodium perborate monohydrate
Sodium perborate monohydrate
Formula: BH₂NaO₄·1H₂O
MW: 117.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149230
CAS Number: 10332-33-9
EINECS: 239-172-9
UN: 3377
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR