Calcium metasilicate ≥99%

Synonyms: Calcium silicate

372668-250G 372668-50G
220061-790EA 541.54 USD
220061-790 220061-791
Calcium metasilicate ≥99%
Calcium metasilicate
Formula: CaO₃Si
MW: 116.16 g/mol
MDL Number: MFCD00015979
CAS Number: 10101-39-0
EINECS: 233-250-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR