Lead(II) sulfide ≥99.9%

Danger

Synonyms: Lead sulphide

372595-100G 372595-25G
220061-785EA 495.38 USD
220061-785 220061-786
Lead(II) sulfide ≥99.9%
Lead(II) sulphide
Formula: ̍PbS
MW: 239.27 g/mol
Boiling Pt: 1344 °C (1013 hPa)
Melting Pt: 1114 °C
Density: 7.5 g/cm³ (20 °C)
MDL Number: MFCD00016280
CAS Number: 1314-87-0
EINECS: 215-246-6
UN: 3077
ADR: 9,III
Merck Index: 12,05445

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR