1,3,5-Trimethylbenzene-[D12]

Synonyms: Mesitylene-D12

372374-1G 372374-5G
220061-775EA 269.23 USD
220061-775 220061-776
1,3,5-Trimethylbenzene-[D12]
1,3,5-Trimethylbenzene-[D12]
Formula: C₆D₃(CD₃)₃
MW: 132.1 g/mol
CAS Number: 69441-16-3
UN: 2325
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR