Methyl pyruvate ≥90%, Technical Grade

Synonyms: Pyruvic acid ethyl mester, 2-Ketopropionic acid methyl ester, 2-Oxopropionic acid methyl ester, Methyl 2-oxopropanoate, α-Ketopropionic acid methyl ester

371173-100G 371173-25G
220061-724EA 117.38 USD
220061-724 220061-725
Methyl pyruvate ≥90%, Technical Grade
Methyl pyruvate
Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 137 °C (1013 hPa)
Density: 1.154 g/cm³ (0 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008754
CAS Number: 600-22-6
EINECS: 209-987-4
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR