α-Bromo-4-(trifluoromethoxy)toluene ≥97%

Danger

370622-1G 370622-5G
220061-694EA 87.23 USD
220061-694 220061-695
α-Bromo-4-(trifluoromethoxy)toluene ≥97%
α-Bromo-4-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 23…24 °C
Density: 1.58 g/cm³ (20 °C)
MDL Number: MFCD00061238
CAS Number: 50824-05-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR