1H,1H,2H-Perfluoro-1-decene ≥99%

Warning

370576-25G 370576-5G
220061-643EA 94.98 USD
220061-643 220061-644
1H,1H,2H-Perfluoro-1-decene ≥99%
1H,1H,2H-Perfluoro-1-decene
Formula: C₁₀H₃F₁₇
MW: 446.11 g/mol
Boiling Pt: 145…150 °C (1013 hPa)
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039246
CAS Number: 21652-58-4
EINECS: 244-503-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR