1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol

370533-5G 370533-25G
220061-214EA 54.82 USD
220061-214 220061-213
1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%
1H,1H,2H,2H-Tridecafluoro-1-octanol
Formula: C₈H₅F₁₃O
MW: 364.11 g/mol
MDL Number: MFCD00042143
CAS Number: 647-42-7
EINECS: 211-477-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR