1H,1H,2H,2H-Perfluoro-1-iodooctane ≥96%

Warning

Synonyms: 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 1H,1H,2H,2H-Perfluorooctyl iodide, 1-Iodo-3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctane

370517-25G 370517-5G 370517-100G
220061-210EA 143.23 USD
220061-210 220061-211 220061-209
1H,1H,2H,2H-Perfluoro-1-iodooctane ≥96%
1H,1H,2H,2H-Perfluoro-1-iodooctane
Formula: C₈H₄F₁₃I
MW: 474 g/mol
Boiling Pt: 179 °C (1013 hPa)
Melting Pt: 20.5 °C
Density: 1.91 g/cm³ (25 °C)
MDL Number: MFCD00039410
CAS Number: 2043-57-4
EINECS: 218-056-1

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR