4-Nitro-3-picoline-N-oxide

Synonyms: 4-Nitropicoline N-oxide, 3-Methyl-4-nitropyridine 1-oxide, 3-Methyl-4-nitropyridine-N-oxide, 3-Methyl-4-nitropyridine N-oxide, 4-Nitro-3-picoline N-oxide, 3-Methyl-4-nitropyridine-1-oxide

370061-5G 370061-1G
220061-632EA 513.8 USD
220061-632 220061-631
4-Nitro-3-picoline-N-oxide
4-Nitro-3-picoline-N-oxide
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 137.5 °C
MDL Number: MFCD00014626
CAS Number: 1074-98-2
EINECS: 214-050-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=295d4261ff1b92687eee57e12c17849e4f5b801e&vid=1d97a39ea83235463da9c47fa3b187266470e7fa&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR