1,3-Difluoro-4-iodobenzene ≥98%

Warning

Synonyms: 2,4-Difluoroiodobenzene, 2,4-Difluoro-1-iodobenzene

369101-25G 369101-5G
220061-538EA 616.15 USD
220061-538 220061-539
1,3-Difluoro-4-iodobenzene ≥98%
1,3-Difluoro-4-iodobenzene
Formula: C₆H₃F₂I
MW: 239.99 g/mol
Boiling Pt: 175…176 °C (1013 hPa)
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00040151
CAS Number: 2265-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR