5-Fluoroanthranilic acid ≥97%

Warning

Synonyms: 2-Amino-5-fluorobenzoic acid

367982-10G 367982-1G
220060-674EA 474.92 USD
220060-674 220060-675
5-Fluoroanthranilic acid ≥97%
5-Fluoroanthranilic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00055566
CAS Number: 446-08-2
EINECS: 207-159-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR