Sodium trifluoromethanesulfonate ≥98%

Synonyms: Trifluoromethanesulphonic acid sodium salt

367907-25G 367907-5G
220060-666EA 145.38 USD
220060-666 220060-667
Sodium trifluoromethanesulfonate ≥98%
Sodium trifluoromethanesulphonate
Formula: CF₃NaO₃S
MW: 172.06 g/mol
MDL Number: MFCD00061607
CAS Number: 2926-30-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR