Oleoyl chloride ≥85%, Technical Grade

Danger

Synonyms: Oleic acid chloride

367850-500ML 367850-100ML
220061-473EA 314.46 USD
220061-473 220061-472
Oleoyl chloride ≥85%, Technical Grade
Oleoyl chloride
Formula: C₁₈H₃₃ClO
MW: 300.91 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: –46 °C
Density: 0.9147 g/cm³ (20 °C)
Flash Pt: 171 °C
Storage Temperature: Ambient
MDL Number: MFCD00134332
CAS Number: 112-77-6
EINECS: 204-005-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR