Cerium triacetate hydrate ≥99.9%

Synonyms: Acetic acid cerium (III) salt hydrate, Cerium (III) acetate hydrate

367753-50G 367753-250G
220061-461EA 106.31 USD
220061-461 220061-460
Cerium triacetate hydrate ≥99.9%
Cerium triacetate hydrate
Formula: C₆H₉CeO₆·H₂O
MW: 335.266 g/mol
MDL Number: MFCD00150533
CAS Number: 206996-60-3
EINECS: 208-654-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR