2,4,6-Trihydroxybenzoic acid monohydrate ≥90%, Technical Grade

Synonyms: 2,4,6-Trihydroxybenzoic acid hydrate

367346-100G 367346-25G
220061-389EA 368.77 USD
220061-389 220061-390
2,4,6-Trihydroxybenzoic acid monohydrate ≥90%, Technical Grade
2,4,6-Trihydroxybenzoic acid monohydrate
Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149095
CAS Number: 71989-93-0
EINECS: 201-467-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR