Acetonitrile-[D₃]

Danger

Synonyms: Cyanomethane-D3, Acetonitrile-d3, Ethannitrile-D3, Trideuteroacetonitrile, Methyl-d3 cyanide, Methyl cyanide-D3, Acetonitrile-[²H₃]

366544-25G 366544-50G 366544-5G
220061-217EA 69.54 USD
220061-217 220061-218 220061-219
Acetonitrile-[D₃]
Acetonitrile-[D3]
Formula: D₃CCN
MW: 44.03 g/mol
Boiling Pt: 80.7 °C (1013 hPa)
Melting Pt: -42 °C
Density: 0.844 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
EINECS: 218-616-5
UN: 1648
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR