2-Chloro-5-(trifluoromethyl)pyridine

Synonyms: 2-Chloro-5-trifluoromethylpyridine

366137-50G 366137-10G
220061-136EA 138.92 USD
220061-136 220061-135
2-Chloro-5-(trifluoromethyl)pyridine
2-Chloro-5-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: 30…31 °C
Density: 1.41 g/cm³ (20 °C)
MDL Number: MFCD00042225
CAS Number: 52334-81-3
EINECS: 257-856-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR