2-Amino-4-(trifluoromethyl)thiophenol hydrochloride ≥97%

Synonyms: 2-Amino-4-(trifluoromethyl)benzenethiol hydrochloride

365734-25G 365734-5G
220061-095EA 299.38 USD
220061-095 220061-096
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride ≥97%
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride
Formula: C₇H₇ClF₃NS
MDL Number: MFCD00042600
CAS Number: 4274-38-8
EINECS: 429-560-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR