1-Bromo-2-nitrobenzene ≥98%

Synonyms: 2-Bromonitrobenzene

365424-25G 365424-100G
220061-059EA 337.08 USD
220061-059 220061-058
1-Bromo-2-nitrobenzene ≥98%
1-Bromo-2-nitrobenzene
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 39 - 42 °C
Density: 1.62 g/cm³ (20 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007045
CAS Number: 577-19-5
EINECS: 209-409-0
UN: 3459
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR