3-Fluoro-4-methoxybenzoic acid ≥98%

365114-1G 365114-5G
220061-020EA 50.72 USD
220061-020 220061-021
3-Fluoro-4-methoxybenzoic acid ≥98%
3-Fluoro-4-methoxybenzoic acid
Formula: C₈H₇FO₃
MW: 170.14 g/mol
Melting Pt: 211.5 °C
MDL Number: MFCD00060675
CAS Number: 403-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR