2-Bromo-5-fluorotoluene ≥98%

Danger

Synonyms: 1-Bromo-4-fluoro-2-methylbenzene

365009-5G 365009-25G
220061-009EA 30.8 USD
220061-009 220061-008
2-Bromo-5-fluorotoluene ≥98%
2-Bromo-5-fluorotoluene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 177 °C (1013 hPa)
Density: 1.49 g/cm³ (20 °C)
MDL Number: MFCD00017921
CAS Number: 452-63-1
EINECS: 207-203-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR