Ethyl propenyl ether cis- and trans-mixture ≥98%

Synonyms: Ethyl 1-Propenyl ether

364045-10G 364045-25G
220060-034EA 116.51 USD
220060-034 220060-035
Ethyl propenyl ether cis- and trans-mixture ≥98%
Ethyl propenyl ether
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 55 - 65 °C (1013 hPa)
Density: 0.7951 g/cm³ (0 °C)
MDL Number: MFCD00015184
CAS Number: 928-55-2
EINECS: 213-176-0
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR