(1R,2R)-(+)-1,2-Diphenylethylenediamine ≥97%

Synonyms: (1R,2R)-(+)-1,2-Diphenylethylenediamine, (1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, (1R,2R)-1,2-Diphenylethane-1,2-diamine, (R,R)-DPEN, (+)-((R,R)-1,2-Diamino-2-phenylethyl)benzene

364010-1G 364010-500MG
220060-529EA 112 USD
220060-529 220060-032
(1R,2R)-(+)-1,2-Diphenylethylenediamine ≥97%
(1R,2R)-(+)-1,2-Diphenylethylenediamine
Formula: C₁₄H₁₆N₂
MW: 212.29 g/mol
MDL Number: MFCD00082769
CAS Number: 35132-20-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR