Ethyl-4-bromobenzoate ≥98%

Synonyms: 4-Bromobenzoic acid ethyl ester, Ethyl 4-bromobenzoate

363774-100G 363774-25G
220060-462EA 180.92 USD
220060-462 220060-463
Ethyl-4-bromobenzoate ≥98%
Ethyl-4-bromobenzoate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 18 °C
Density: 1.4332 g/cm³ (17.3 °C)
MDL Number: MFCD00016329
CAS Number: 5798-75-4
EINECS: 227-347-2
Merck Index: 13,01392

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR