2,3-Dichloro-1-propanol ≥97.0%

36295-25ML 36295-5ML
220060-907EA 260.62 USD
220060-907 220060-908
2,3-Dichloro-1-propanol ≥97.0%
2,3-Dichloro-1-propanol
Formula: C₃H₆Cl₂O
MW: 128.99 g/mol
Boiling Pt: 183 °C (1013 hPa)
Melting Pt: -44 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040446
CAS Number: 616-23-9
EINECS: 210-470-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR