2,5-Dimethyl-1,5-hexadiene

Synonyms: 2,5-Dimethylhexa-1,5-diene

362247-10ML 362247-1ML
220060-879EA 519.38 USD
220060-879 220060-880
2,5-Dimethyl-1,5-hexadiene
2,5-Dimethyl-1,5-hexadiene
Formula: C₈H₁₄
MW: 110.2 g/mol
Boiling Pt: 114.3 °C (1013 hPa)
Melting Pt: –75.6 °C
Density: 0.743 g/cm³ (20 °C)
MDL Number: MFCD00008602
CAS Number: 627-58-7
EINECS: 211-003-3
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR