cis-1,2-Cyclopentanediol ≥98%

361437-1G 361437-5G
220060-485EA 70.88 USD
220060-485 220060-486
cis-1,2-Cyclopentanediol ≥98%
cis-1,2-Cyclopentanediol
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: 30 °C
MDL Number: MFCD00134192
CAS Number: 5057-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR