Tetrabutylammonium fluoride ≥75% (w/w) in aqueous solution

361399-500G 361399-100G 361399-25G
220058-912EA 339.23 USD
220058-912 220058-910 220058-911
Tetrabutylammonium fluoride ≥75% (w/w) in aqueous solution
Tetrabutylammonium fluoride
Formula: C₁₆H₃₆FN
MW: 261.47 g/mol
MDL Number: MFCD00011747
CAS Number: 429-41-4
EINECS: 207-057-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR