4-Chloro-3-nitrophenol ≥98%

Warning

361127-5G 361127-25G
220060-777EA 70.22 USD
220060-777 220060-776
4-Chloro-3-nitrophenol ≥98%
4-Chloro-3-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
Melting Pt: 125 - 128 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00043546
CAS Number: 610-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR