2-Phenylbenzyl bromide ≥97%

Synonyms: o-(Bromomethyl)biphenyl, 2-(Bromomethyl)biphenyl, 2-Phenylbenzylbromide

360082-5G 360082-25G
220060-616EA 95.38 USD
220060-616 220060-615
2-Phenylbenzyl bromide ≥97%
2-Phenylbenzyl bromide
Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
MDL Number: MFCD00075454
CAS Number: 19853-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR