Calcium citrate tetrahydrate ≥99%

Synonyms: tri-Calcium dicitrate tetrahydrate, Calcium citrate tribasic tetrahydrate

359734-1KG 359734-250G
220060-441EA 107.58 USD
220060-441 220060-442
Calcium citrate tetrahydrate ≥99%
Calcium citrate tetrahydrate
Formula: C₁₂H₁₀Ca₃O₁₄·4H₂O
MW: 570.5 g/mol
MDL Number: MFCD00150786
CAS Number: 5785-44-4
EINECS: 212-391-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR