Methyl (methylthio)acetate ≥98%

Synonyms: (Methylthio)acetic methyl ester

359440-10ML 359440-50ML
220060-417EA 650.77 USD
220060-417 220060-418
Methyl (methylthio)acetate ≥98%
Methyl (methylthio)acetate
Formula: C₄H₈O₂S
MW: 120.17 g/mol
Boiling Pt: 160…161 °C (1013 hPa)
Density: 1.121 g/cm³ (20 °C)
MDL Number: MFCD00039809
CAS Number: 16630-66-3
EINECS: 240-683-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR