Perfluorooctane ≥98%

Synonyms: Octadecafluorooctane

359238-100G 359238-25G
220060-392EA 242.31 USD
220060-392 220060-393
Perfluorooctane ≥98%
Perfluorooctane
Formula: C₈F₁₈
MW: 438.06 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: –25 °C
Density: 1.73 g/cm³ (20 °C)
MDL Number: MFCD00042083
CAS Number: 307-34-6
EINECS: 206-199-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR