Disodium hydrogen citrate sesquihydrate ≥99%

Synonyms: Citric acid disodium salt sesquihydrate, Sodium hydrogencitrate sesquihydrate, Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrate, Sodium citrate dibasic sesquihydrate

359084-1KG 359084-250G
220060-381EA 120.62 USD
220060-381 220060-382
Disodium hydrogen citrate sesquihydrate ≥99%
di-Sodium hydrogen citrate sesquihydrate
Formula: C₆H₆Na₂O₇·1.5H₂O
MW: 263.11 g/mol
MDL Number: MFCD00150445
CAS Number: 6132-05-4
EINECS: 242-734-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR