Cerium(IV) sulfate

Warning

359009-25G 359009-100G
220059-707EA 32.18 USD
220059-707 220059-706
Cerium(IV) sulfate
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
Melting Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3260
ADR: 8,III
Merck Index: 12,02036

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR