1,1,2,2-Tetrachloroethane-[D₂]

Danger

Synonyms: 1,1,2,2-Tetrachloroethane-[²H₂]

358703-25G 358703-1G 358703-5G
220060-367EA 74.15 USD
220060-367 220060-366 220060-368
1,1,2,2-Tetrachloroethane-[D₂]
1,1,2,2-Tetrachloroethane-[D₂]
Formula: Cl₂CDCDCl₂
MW: 169.83 g/mol
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: –43 °C
Density: 1.6118 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00037672
CAS Number: 33685-54-0
EINECS: 251-634-1
UN: 1702
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR