Tetrabutylammonium cyanide

Danger

358665-1G 358665-5G
220060-360EA 39.66 USD
220060-360 220060-361
Tetrabutylammonium cyanide
Tetrabutylammonium cyanide
Formula: C₁₇H₃₆N₂
MW: 268.49 g/mol
Storage Temperature: Ambient
CAS Number: 10442-39-4
EINECS: 233-926-0
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR