4'-Iodoacetophenone ≥98%

Warning

Synonyms: 1-(4-Iodophenyl)ethan-1-one, p-Iodoacetophenone, 1-(4-Iodophenyl)ethanone

357804-5G 357804-25G
220060-326EA 43.08 USD
220060-326 220060-325
4'-Iodoacetophenone ≥98%
4'-Iodoacetophenone
Formula: C₈H₇IO
MW: 246.05 g/mol
Boiling Pt: 145 - 150 °C (20 -rr)
Melting Pt: 80 - 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00045320
CAS Number: 13329-40-3
EINECS: 236-372-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR