(2-Methoxyethoxy)methyl chloride, Technical Grade

Synonyms: 1-Chloromethoxy-2-methoxyethane, 1-Chloro-2,5-dioxahexane, MEM chloride

357480-25G 357480-5G
220059-649EA 200.31 USD
220059-649 220060-291
(2-Methoxyethoxy)methyl chloride, Technical Grade
(2-Methoxyethoxy)methyl chloride
Formula: ClCH₂OCH₂CH₂OCH₃
MW: 124.57 g/mol
Boiling Pt: 50 °C (1013 hPa)
Density: 1.094 g/cm³ (20 °C)
MDL Number: MFCD00000888
CAS Number: 3970-21-6
EINECS: 223-589-8
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR