2-Bromo-4-methylpyridine

349984-10G 349984-1G
220059-675EA 140 USD
220059-675 220059-676
2-Bromo-4-methylpyridine
2-Bromo-4-methylpyridine
Formula: C₆H₆BrN
Boiling Pt: 42…44 °C (0.6 mmHg)
Melting Pt: 218…228 °C
Density: 1.545 g/cm³ (20 °C)
MDL Number: MFCD00082590
CAS Number: 4926-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR